Gene Transcripts

Gene Symbol
IL17B
Entrez Gene ID
27190
Gene Name
interleukin 17B
Gene Aliases
IL-17B, IL-20, NIRF, ZCYTO7
Chromosome Location
Chr. 5: 149374267 - 149404202 on Build GRCh38
UniGene ID
Hs.156979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014443.2 NP_055258.1 1, 2 75
GenBank mRNA AF110385.1 - 1, 2 68
AF152098.1 - 1, 2 66
AF184969.1 - 1, 2 66
AF212311.1 - 1, 2 75
AF218727.1 - 1, 2 66
AY358931.1 - 1, 2 66
BC113925.1 - 1, 2 48 40084832
BC113946.1 - 1, 2 48 40084836
CR457192.1 - 1, 2 25

View Full Product Details