Gene Transcripts
- Gene Symbol
- ERLEC1
- Entrez Gene ID
- 27248
- Gene Name
- endoplasmic reticulum lectin 1
- Gene Aliases
- C2orf30, CIM, CL24936, CL25084, HEL117, XTP3-B, XTP3TPB
- Chromosome Location
- Chr. 2: 53786931 - 53826928 on Build GRCh38
- UniGene ID
- Hs.438336
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127397.2 |
NP_001120869.1
|
9, 10 |
1331 |
|
NM_001127398.2 |
NP_001120870.1
|
8, 9 |
1169 |
|
NM_015701.4 |
NP_056516.2
|
9, 10 |
1331 |
|
XM_011532766.1 |
XP_011531068.1
|
9, 10 |
1245 |
|
GenBank mRNA |
AF131743.1 |
- |
1, 2 |
150 |
|
AF131849.1 |
- |
3, 4 |
520 |
|
AK001913.1 |
- |
9, 10 |
1212 |
|
AK315477.1 |
- |
9, 10 |
1120 |
|
AY358717.1 |
- |
8, 9 |
1053 |
|
AY453410.1 |
- |
9, 10 |
1182 |
|
BC013129.1 |
- |
9, 10 |
1182 |
3447840 |
BC022228.1 |
- |
8, 9 |
1017 |
4447432 |
BQ017438.1 |
- |
4 |
362 |
|
EU794676.1 |
- |
9, 10 |
1331 |
|
View Full Product Details