Gene Transcripts
- Gene Symbol
- GLS
- Entrez Gene ID
- 2744
- Gene Name
- glutaminase
- Gene Aliases
- AAD20, GAC, GAM, GLS1, KGA
- Chromosome Location
- Chr. 2: 190880821 - 190965552 on Build GRCh38
- UniGene ID
- Hs.116448
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256310.1 |
NP_001243239.1
|
9, 10 |
1399 |
|
NM_014905.4 |
NP_055720.3
|
9, 10 |
1399 |
|
XM_005246467.3 |
XP_005246524.1
|
9, 10 |
1399 |
|
XM_006712435.1 |
XP_006712498.1
|
9, 10 |
1399 |
|
GenBank mRNA |
AB020645.1 |
- |
9, 10 |
1385 |
|
AF097492.1 |
- |
9, 10 |
1033 |
|
AF097493.1 |
- |
2, 3 |
127 |
|
AF158555.1 |
- |
10, 11 |
1379 |
|
AF223943.1 |
- |
9, 10 |
1154 |
|
AF279697.1 |
- |
6, 7 |
484 |
|
AF327434.1 |
- |
9, 10 |
1365 |
|
AK001220.1 |
- |
6, 7 |
471 |
|
AK289747.1 |
- |
10, 11 |
1375 |
|
BC038507.1 |
- |
10, 11 |
1375 |
5263220 |
CR749593.1 |
- |
2, 3 |
1858 |
|
View Full Product Details