Gene Transcripts

Gene Symbol
FBXL22
Entrez Gene ID
283807
Gene Name
F-box and leucine rich repeat protein 22
Gene Aliases
Fbl22
Chromosome Location
Chr. 15: 63596225 - 63608898 on Build GRCh38
UniGene ID
Hs.631163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203373.2 NP_976307.2 1 127
XM_005254320.3 XP_005254377.1 1 1255
XM_011521468.2 XP_011519770.1 1 93
XM_011521469.1 XP_011519771.1 1 89
XM_011521472.2 XP_011519774.1 1 93
XM_017022086.1 XP_016877575.1 1 1255
GenBank mRNA AA495872.1 - 1 127
AK096945.1 - 1 89
BC065833.1 - 1 83 30390268

View Full Product Details