Gene Transcripts

Gene Symbol
BTBD8
Entrez Gene ID
284697
Gene Name
BTB domain containing 8
Gene Aliases
Chromosome Location
Chr. 1: 92080305 - 92147840 on Build GRCh38
UniGene ID
Hs.676102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183242.3 NP_899065.2 6 1032
GenBank mRNA AK296516.1 - 6 799
AY346333.1 - 6 1033
BC013922.1 - 6 1005 4048340

View Full Product Details