Gene Transcripts

Gene Symbol
GPS1
Entrez Gene ID
2873
Gene Name
G protein pathway suppressor 1
Gene Aliases
COPS1, CSN1, SGN1
Chromosome Location
Chr. 17: 82051477 - 82057470 on Build GRCh38
UniGene ID
Hs.268530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321089.1 NP_001308018.1 6 908
NM_001321090.1 NP_001308019.1 6 896
NM_001321091.1 NP_001308020.1 6 896
NM_001321092.1 NP_001308021.1 5 692
NM_001321093.1 NP_001308022.1 6 777
NM_004127.5 NP_004118.3 5 704
NM_212492.2 NP_997657.1 5 1187
XM_005256350.1 XP_005256407.1 6 876
XM_005256353.1 XP_005256410.1 6 864
XM_005256359.1 XP_005256416.1 5 668
XM_005256360.3 XP_005256417.1 5 697
XM_011523569.1 XP_011521871.1 6 729
XM_011523570.1 XP_011521872.1 6 767
XM_017024524.1 XP_016880013.1 5 686
XM_017024525.1 XP_016880014.1 6 864
XM_017024526.1 XP_016880015.1 5 686
XM_017024527.1 XP_016880016.1 5 697
XM_017024528.1 XP_016880017.1 5 652
XM_017024529.1 XP_016880018.1 5 652
XM_017024530.1 XP_016880019.1 5 640
XM_017024531.1 XP_016880020.1 6 717
XM_017024532.1 XP_016880021.1 5 668
XM_017024533.1 XP_016880022.1 5 684
XM_017024534.1 XP_016880023.1 6 729
XM_017024535.1 XP_016880024.1 6 717
GenBank mRNA AB209596.1 - 3 1756
AK026610.1 - 6 739
AK093283.1 - 5 622
AK222508.1 - 5 657
AK225248.1 - 5 654
AK289435.1 - 5 810
AK297872.1 - 5 680
BC000155.2 - 5 660 2900944
BC064503.1 - 5 1099 6572943
BT009834.1 - 4 589
U20285.2 - 5 684

View Full Product Details