Gene Transcripts

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
Chromosome Location
Chr. 15: 72343435 - 72376473 on Build GRCh38
UniGene ID
Hs.604479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000520.5 NP_000511.2 6 1089
NM_001318825.1 NP_001305754.1 6 1122
NR_134869.1 6 1089
GenBank mRNA AK222502.1 - 6 602
AK296528.1 - 6 460
AK297678.1 - 5 521
AK301000.1 - 6 642
AK301138.1 - 4 461
AK307770.1 - 6 609
AK310154.1 - 5 572
BC001138.2 - 4 229
BC018927.2 - 6 599 3353424
BC034424.1 - 3 2568
BC084537.1 - 6 595 5459166
BX537989.1 - 5 5915
CR627386.1 - 6 1089
KU178109.1 - 6 588
M13520.1 - 6 472

View Full Product Details