Gene Transcripts

Gene Symbol
UBE2K
Entrez Gene ID
3093
Gene Name
ubiquitin conjugating enzyme E2 K
Gene Aliases
E2-25K, HIP2, HYPG, LIG, UBC1
Chromosome Location
Chr. 4: 39698044 - 39782792 on Build GRCh38
UniGene ID
Hs.50308
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111112.1 NP_001104582.1 6 736
NM_001111113.1 NP_001104583.1 5 712
NM_001312646.1 NP_001299575.1 5 682
NM_001312647.1 NP_001299576.1 7 777
NM_001312648.1 NP_001299577.1 6 771
NM_005339.4 NP_005330.1 7 865
GenBank mRNA AB022435.1 - 7 667
AB022436.1 - 5 514
AK093533.1 - 7 686
AK291454.1 - 5 617
AK295846.1 - 5 617
AK307130.1 - 6 676
AK315524.1 - 7 791
BC022804.1 - 7 741 4893778
BC050600.1 - 7 719 6502901
CX786829.1 - 6 575
U58522.1 - 7 584

View Full Product Details