Gene Transcripts

Gene Symbol
SNAI3
Entrez Gene ID
333929
Gene Name
snail family transcriptional repressor 3
Gene Aliases
SMUC, SNAIL3, ZNF293, Zfp293
Chromosome Location
Chr. 16: 88677682 - 88686474 on Build GRCh38
UniGene ID
Hs.673548
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178310.3 NP_840101.1 1, 2 150
GenBank mRNA BC041461.1 - 1, 2 57
BC107058.2 - 1, 2 150 40005947

View Full Product Details