Gene Transcripts

Gene Symbol
SLCO1B7
Entrez Gene ID
338821
Gene Name
solute carrier organic anion transporter family member 1B7 (putative)
Gene Aliases
LST-3, LST-3TM12, LST3, SLC21A21
Chromosome Location
Chr. 12: 21015696 - 21090106 on Build GRCh38
UniGene ID
Hs.729117
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009562.4 NP_001009562.3 10 1490
GenBank mRNA AF401642.1 - 10 1490

View Full Product Details