Gene Transcripts
- Gene Symbol
- MYBPHL
- Entrez Gene ID
- 343263
- Gene Name
- myosin binding protein H like
- Gene Aliases
- Chromosome Location
- Chr. 1: 109292365 - 109309394 on Build GRCh38
- UniGene ID
- Hs.443402
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001010985.2 |
NP_001010985.2
|
7, 8 |
1101 |
|
NM_001265613.1 |
NP_001252542.1
|
7, 8 |
1032 |
|
XM_017001173.1 |
XP_016856662.1
|
7, 8 |
3454 |
|
XM_017001174.1 |
XP_016856663.1
|
7, 8 |
3454 |
|
XM_017001175.1 |
XP_016856664.1
|
7, 8 |
3385 |
|
GenBank mRNA |
AK129834.1 |
- |
7, 8 |
1101 |
|
BC108664.1 |
- |
2, 3 |
334 |
|
BC132900.1 |
- |
7, 8 |
1078 |
40146922 |
BC132902.1 |
- |
7, 8 |
1078 |
40146924 |
BC144471.1 |
- |
7, 8 |
1009 |
9052998 |
View Full Product Details