Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000641.3 | NP_000632.1 | 2 | 193 | |
GenBank mRNA | AK290572.1 | - | 2 | 193 | |
AK315294.1 | - | 2 | 193 | ||
BC012506.1 | - | 2 | 176 | 4476678 | |
M57765.1 | - | 2 | 176 | ||
X58377.1 | - | 2 | 103 |