Gene Transcripts
- Gene Symbol
- INSM1
- Entrez Gene ID
- 3642
- Gene Name
- INSM transcriptional repressor 1
- Gene Aliases
- IA-1, IA1
- Chromosome Location
- Chr. 20: 20368121 - 20370949 on Build GRCh38
- UniGene ID
- Hs.89584
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002196.2 |
NP_002187.1
|
1 |
1259 |
|
GenBank mRNA |
M93119.1 |
- |
1 |
1259 |
|
View Full Product Details