Gene Transcripts
- Gene Symbol
- DNAJB13
- Entrez Gene ID
- 374407
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B13
- Gene Aliases
- RSPH16A, TSARG5, TSARG6
- Chromosome Location
- Chr. 11: 73950319 - 73970287 on Build GRCh38
- UniGene ID
- Hs.567888
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153614.3 |
NP_705842.2
|
2, 3 |
933 |
|
XM_011545004.2 |
XP_011543306.1
|
4, 5 |
1184 |
|
XM_011545007.2 |
XP_011543309.1
|
2, 3 |
709 |
|
XM_011545013.2 |
XP_011543315.1
|
4, 5 |
1190 |
|
XM_011545014.2 |
XP_011543316.1
|
4, 5 |
1190 |
|
XM_017017675.1 |
XP_016873164.1
|
3, 4 |
454 |
|
XM_017017679.1 |
XP_016873168.1
|
4, 5 |
1190 |
|
GenBank mRNA |
AK128589.1 |
- |
2, 3 |
226 |
|
AY138810.2 |
- |
2, 3 |
933 |
|
AY325766.1 |
- |
2, 3 |
226 |
|
View Full Product Details