Gene Transcripts
- Gene Symbol
- C19orf35
- Entrez Gene ID
- 374872
- Gene Name
- chromosome 19 open reading frame 35
- Gene Aliases
- Chromosome Location
- Chr. 19: 2274632 - 2282182 on Build GRCh38
- UniGene ID
- Hs.511803
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198532.2 |
NP_940934.1
|
2, 3 |
175 |
|
GenBank mRNA |
AK127680.1 |
- |
2, 3 |
169 |
|
View Full Product Details