Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002274.3 | NP_002265.2 | 5 | 982 | |
NM_153490.2 | NP_705694.2 | 5 | 982 | ||
GenBank mRNA | AK092276.1 | - | 6 | 947 | |
AK223051.1 | - | 5 | 981 | ||
AK223077.1 | - | 5 | 981 | ||
AK290244.1 | - | 5 | 981 | ||
AK296806.1 | - | 6 | 770 | ||
AK309752.1 | - | 5 | 983 | ||
BC002661.2 | - | 5 | 966 | 3610361 | |
BC077718.1 | - | 5 | 959 | 5479174 | |
BC126184.1 | - | 5 | 946 | 8991900 | |
BC136494.1 | - | 5 | 946 | 9020483 | |
DR422220.1 | - | 5 | 465 | ||
X14640.1 | - | 5 | 961 | ||
X52426.1 | - | 5 | 966 |