Gene Transcripts

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
Chromosome Location
Chr. 14: 73478484 - 73634418 on Build GRCh38
UniGene ID
Hs.660774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001220484.1 NP_001207413.1 14 2773
NM_203309.2 NP_976054.2 13 2694
XM_006720143.2 XP_006720206.1 13 2731
XM_011536760.2 XP_011535062.1 13 2917
XM_017021289.1 XP_016876778.1 13 3255
GenBank mRNA AK302564.1 - 14 2747
BC047590.1 - 13 2694 4837558
GU727638.1 - 12 2284

View Full Product Details