Gene Transcripts
- Gene Symbol
- NOTCH2
- Entrez Gene ID
- 4853
- Gene Name
- notch 2
- Gene Aliases
- AGS2, HJCYS, hN2
- Chromosome Location
- Chr. 1: 119911553 - 120073449 on Build GRCh38
- UniGene ID
- Hs.487360
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001200001.1 |
NP_001186930.1
|
15, 16 |
2784 |
|
NM_024408.3 |
NP_077719.2
|
15, 16 |
2784 |
|
XM_005270901.3 |
XP_005270958.1
|
16, 17 |
5098 |
|
XM_011541519.2 |
XP_011539821.1
|
15, 16 |
7892 |
|
XM_011541520.2 |
XP_011539822.1
|
15, 16 |
4802 |
|
XM_017001372.1 |
XP_016856861.1
|
15, 16 |
2454 |
|
XM_017001373.1 |
XP_016856862.1
|
15, 16 |
2433 |
|
GenBank mRNA |
AF308601.1 |
- |
15, 16 |
2499 |
|
AF315356.1 |
- |
15, 16 |
2499 |
|
BC071562.1 |
- |
15, 16 |
2733 |
4822955 |
View Full Product Details