Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016125.3 | NP_057209.3 | 6 | 1083 | |
GenBank mRNA | AK098649.1 | - | 4 | 463 | |
AK296197.1 | - | 4 | 886 | ||
BC006971.1 | - | 5 | 960 | ||
BC088365.1 | - | 6 | 1046 | 30390062 |