Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079520.1 | NP_001072988.1 | 3 | 714 | |
NM_016651.5 | NP_057735.2 | 3 | 714 | ||
NR_046093.1 | 3 | 323 | |||
NR_046095.1 | 3 | 631 | |||
XM_006720167.3 | XP_006720230.1 | 3 | 651 | ||
GenBank mRNA | AK299879.1 | - | 3 | 631 | |
AK316164.1 | - | 3 | 421 | ||
AK316169.1 | - | 1 | 37 | ||
AK316237.1 | - | 3 | 323 | ||
AK316423.1 | - | 2 | 190 | ||
DR157190.1 | - | 2 | 92 |