Gene Transcripts

Gene Symbol
DPPA4
Entrez Gene ID
55211
Gene Name
developmental pluripotency associated 4
Gene Aliases
2410091M23Rik
Chromosome Location
Chr. 3: 109326141 - 109337572 on Build GRCh38
UniGene ID
Hs.317659
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018189.3 NP_060659.3 2 143
XM_011512954.2 XP_011511256.1 2 422
GenBank mRNA AK001575.1 - 2 85
AK022698.1 - 2 100
AK290992.1 - 2 136
AK298623.1 - 2 93
AK299290.1 - 2 86
BC032846.2 - 2 117 5267235

View Full Product Details