Gene Transcripts

Gene Symbol
TBC1D19
Entrez Gene ID
55296
Gene Name
TBC1 domain family member 19
Gene Aliases
Chromosome Location
Chr. 4: 26583924 - 26858919 on Build GRCh38
UniGene ID
Hs.479403
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001292054.1 NP_001278983.1 4 532
NM_018317.3 NP_060787.2 7 727
XM_011513852.2 XP_011512154.1 7 567
XM_017008363.1 XP_016863852.1 7 567
XM_017008364.1 XP_016863853.1 7 567
GenBank mRNA AB449905.1 - 4 262
AK001944.1 - 7 567
AK225395.1 - 7 567
AK289631.1 - 7 567
AK310795.1 - 7 574
BC031642.1 - 7 523 5175317

View Full Product Details