Gene Transcripts

Gene Symbol
FAM63A
Entrez Gene ID
55793
Gene Name
family with sequence similarity 63 member A
Gene Aliases
Chromosome Location
Chr. 1: 150996737 - 151008378 on Build GRCh38
UniGene ID
Hs.743952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040217.2 NP_001035307.1 5 991
NM_001163258.1 NP_001156730.1 6 1054
NM_001163259.1 NP_001156731.1 5 956
NM_001163260.1 NP_001156732.1 5 867
NM_001319998.1 NP_001306927.1 6 2371
NM_018379.4 NP_060849.2 6 1655
XM_005245321.3 XP_005245378.1 5 1629
XM_005245325.3 XP_005245382.1 5 984
XM_005245327.3 XP_005245384.1 4 825
XM_005245328.3 XP_005245385.1 4 825
XM_011509757.2 XP_011508059.1 5 1629
XM_017001770.1 XP_016857259.1 5 1629
XM_017001771.1 XP_016857260.1 5 1367
XM_017001772.1 XP_016857261.1 5 1367
XM_017001773.1 XP_016857262.1 5 1629
XM_017001774.1 XP_016857263.1 5 1367
XM_017001777.1 XP_016857266.1 5 981
GenBank mRNA AB037811.1 - 4 840
AK002142.1 - 5 717
AK125493.1 - 5 691
AK125959.1 - 6 2371
AK301946.1 - 6 1054
AK303962.1 - 5 644
BC032321.1 - 5 1131 5240620
BC075812.1 - 6 1642

View Full Product Details