Gene Transcripts

Gene Symbol
PYGM
Entrez Gene ID
5837
Gene Name
phosphorylase, glycogen, muscle
Gene Aliases
Chromosome Location
Chr. 11: 64746389 - 64760715 on Build GRCh38
UniGene ID
Hs.154084
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164716.1 NP_001158188.1 17 2916
GenBank mRNA AF066859.1 - 19 2363
AK056607.1 - 17 2175
AK057547.1 - 2 1715
AK299376.1 - 15 1703
AK299461.1 - 19 2382
AK314474.1 - 19 2367
BC126392.1 - 19 2762 8992108
BC130514.1 - 19 2762 40146545
X03031.1 - 3 340

View Full Product Details