Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031885.3 | NP_114091.3 | 14 | 2016 | |
XM_005256080.2 | XP_005256137.1 | 14 | 2015 | ||
GenBank mRNA | AB208905.1 | - | 9 | 5313 | |
AF342736.1 | - | 14 | 2203 | ||
AK027635.1 | - | 14 | 1944 | ||
AK127290.1 | - | 2 | 2607 | ||
AK289604.1 | - | 14 | 1906 | ||
AL834176.1 | - | 2 | 1364 | ||
BC014140.2 | - | 14 | 1906 | 4653315 |