Gene Transcripts

Gene Symbol
EEFSEC
Entrez Gene ID
60678
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA specific
Gene Aliases
EFSEC, SELB
Chromosome Location
Chr. 3: 128153456 - 128426193 on Build GRCh38
UniGene ID
Hs.477498
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021937.4 NP_068756.2 2 520
XM_005247696.2 XP_005247753.1 2 521
XM_017006975.1 XP_016862464.1 2 477
XM_017006976.1 XP_016862465.1 2 477
XM_017006977.1 XP_016862466.1 2 477
XM_017006978.1 XP_016862467.1 3 300
XM_017006979.1 XP_016862468.1 2 789
XM_017006980.1 XP_016862469.1 2 521
GenBank mRNA AF268872.1 - 2 259
BC007933.2 - 2 398 4120858
BG420135.1 - 2 507

View Full Product Details