Gene Transcripts

Gene Symbol
CCL21
Entrez Gene ID
6366
Gene Name
C-C motif chemokine ligand 21
Gene Aliases
6Ckine, CKb9, ECL, SCYA21, SLC, TCA4
Chromosome Location
Chr. 9: 34709005 - 34710167 on Build GRCh38
UniGene ID
Hs.57907
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002989.3 NP_002980.1 1 119
XM_011518004.2 XP_011516306.1 1 106
GenBank mRNA AB002409.1 - 1 76
AF001979.1 - 1 18
AK311901.1 - 1 88
AY358887.1 - 1 23
BC027918.1 - 1 25 5226781
BI833188.1 - 1 102
BP331359.1 - 1 119
CR450326.1 - 1 18
U88320.1 - 1 32

View Full Product Details