Gene Transcripts

Gene Symbol
CRLF2
Entrez Gene ID
64109
Gene Name
cytokine receptor-like factor 2
Gene Aliases
CRL2, CRLF2Y, TSLPR
Chromosome Location
Chr. X: 1190437 - 1212762 on Build GRCh38
UniGene ID
Hs.287729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012288.2 NP_001012288.2 6, 7 849
NM_022148.3 NP_071431.2 7, 8 952
NR_110830.1 7, 8 1131
XM_011546181.2 XP_011544483.1 7, 8 987
XM_011546182.2 XP_011544484.1 7, 8 917
GenBank mRNA AB052639.1 - 7, 8 863
AF142570.1 - 7, 8 879
AF338733.1 - 7, 8 863
AK026800.1 - 7, 8 1133
AK223303.1 - 7, 8 979
AK225583.1 - 7, 8 1133
AY775789.1 - 6, 7 760

View Full Product Details