Gene Transcripts

Gene Symbol
TRA2B
Entrez Gene ID
6434
Gene Name
transformer 2 beta homolog (Drosophila)
Gene Aliases
Htra2-beta, PPP1R156, SFRS10, SRFS10, TRA2-BETA, TRAN2B
Chromosome Location
Chr. 3: 185914568 - 185938136 on Build GRCh38
UniGene ID
Hs.533122
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243879.1 NP_001230808.1 4 714
NM_004593.2 NP_004584.1 5 848
XM_005247703.1 XP_005247760.1 6 1016
XM_011513072.2 XP_011511374.1 6 3818
XM_017007033.1 XP_016862522.1 5 1994
GenBank mRNA AB209208.1 - 6 3805
AK091863.1 - 1 1735
AK094495.1 - 2 770
AK098191.1 - 5 635
AK125610.1 - 2 1518
AK301118.1 - 4 567
AK308267.1 - 4 592
AK315632.1 - 5 832
BC000160.2 - 5 715 2984771
BC000451.2 - 5 714 2821443
BC005898.1 - 5 699 4046349
CR749214.1 - 4 2875
U61267.1 - 5 693
U68063.1 - 5 701

View Full Product Details