Gene Transcripts

Gene Symbol
FAM83G
Entrez Gene ID
644815
Gene Name
family with sequence similarity 83 member G
Gene Aliases
PAWS1
Chromosome Location
Chr. 17: 18968789 - 19006144 on Build GRCh38
UniGene ID
Hs.710727
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039999.2 NP_001035088.2 5 1190
XM_017024953.1 XP_016880442.1 5 1274
GenBank mRNA AK123558.1 - 3 1460
BX365956.2 - 2 279

View Full Product Details