Gene Transcripts

Gene Symbol
MEIG1
Entrez Gene ID
644890
Gene Name
meiosis/spermiogenesis associated 1
Gene Aliases
SPATA39, bA2K17.3
Chromosome Location
Chr. 10: 14959439 - 14972851 on Build GRCh38
UniGene ID
Hs.257249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080836.2 NP_001074305.1 2, 3 275
GenBank mRNA AA971146.1 - 1, 2 186
BG189536.1 - 1, 2 159

View Full Product Details