Gene Transcripts

Gene Symbol
SPINK8
Entrez Gene ID
646424
Gene Name
serine peptidase inhibitor, Kazal type 8 (putative)
Gene Aliases
Chromosome Location
Chr. 3: 48306846 - 48330311 on Build GRCh38
UniGene ID
Hs.257687
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080525.1 NP_001073994.1 2 96
XM_017007046.1 XP_016862535.1 4 686
XM_017007047.1 XP_016862536.1 4 640

View Full Product Details