Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005631.4 | NP_005622.1 | 4 | 1077 | |
XM_011516522.2 | XP_011514824.1 | 5 | 643 | ||
GenBank mRNA | AF071494.1 | - | 4 | 797 | |
AF120103.1 | - | 4 | 855 | ||
BC009989.2 | - | 4 | 1077 | 4127774 | |
U84401.1 | - | 4 | 797 |