Gene Transcripts

Gene Symbol
ST13
Entrez Gene ID
6767
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein)
Gene Aliases
AAG2, FAM10A1, FAM10A4, HIP, HOP, HSPABP, HSPABP1, P48, PRO0786, SNC6
Chromosome Location
Chr. 22: 40824535 - 40857008 on Build GRCh38
UniGene ID
Hs.712713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278589.1 NP_001265518.1 7 952
NM_003932.4 NP_003923.2 7 982
GenBank mRNA AK130770.1 - 7 615
AK303530.1 - 7 583
AK312399.1 - 7 596
AK316155.1 - 7 501
AY513286.1 - 7 580
AY826824.1 - 7 591
AY826825.1 - 7 591
BC052982.1 - 7 604 6046993
BC071629.1 - 7 582 4536907
BC101206.1 - 7 538
BC101207.2 - 7 538
BC101208.1 - 7 538
BC107148.1 - 7 539 40031944
BC121107.1 - 7 590
BC121108.1 - 7 590
BC139724.1 - 7 539 40031942
BG706239.1 - 661
CR456586.1 - 7 622
U17714.1 - 7 588
U28918.1 - 7 607

Gene Symbol
ST13P4
Entrez Gene ID
145165
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 4
Gene Aliases
FAM10A4, FAM10A4P
Chromosome Location
Chr. 13: 50172018 - 50173616 on Build GRCh38
UniGene ID
Hs.511834
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002183.1 1 573
GenBank mRNA AF539468.1 - 1 573

View Full Product Details