Gene Transcripts

Gene Symbol
TARBP1
Entrez Gene ID
6894
Gene Name
TAR (HIV-1) RNA binding protein 1
Gene Aliases
TRM3, TRP-185, TRP185
Chromosome Location
Chr. 1: 234391313 - 234479174 on Build GRCh38
UniGene ID
Hs.498115
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005646.3 NP_005637.3 14 2247
XM_005273234.3 XP_005273291.1 14 2244
XM_011544263.2 XP_011542565.1 14 2314
XM_017002194.1 XP_016857683.1 14 2317
XM_017002195.1 XP_016857684.1 11 1120
XM_017002196.1 XP_016857685.1 2 92
GenBank mRNA U38847.1 - 14 2247

View Full Product Details