Gene Transcripts

Gene Symbol
TRAF2
Entrez Gene ID
7186
Gene Name
TNF receptor associated factor 2
Gene Aliases
MGC:45012, TRAP, TRAP3
Chromosome Location
Chr. 9: 136881776 - 136926615 on Build GRCh38
UniGene ID
Hs.522506
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021138.3 NP_066961.2 3 293
XM_011518973.2 XP_011517275.1 4 597
XM_011518974.2 XP_011517276.1 3 1198
XM_011518975.2 XP_011517277.1 4 525
XM_011518976.2 XP_011517278.1 4 566
XM_011518977.2 XP_011517279.1 3 286
XM_011518978.2 XP_011517280.1 3 1707
XM_017015094.1 XP_016870583.1 3 341
XM_017015095.1 XP_016870584.1 3 1051
GenBank mRNA AK054686.1 - 3 280
AK222961.1 - 3 290
AK289722.1 - 4 409
AK294607.1 - 3 290
AK298370.1 - 3 257
BC032410.1 - 2 279 5216790
BC033810.1 - 3 281 5182942
BC043492.1 - 3 293 5767794
BC064662.1 - 2 288 6528158
BM785733.1 - 3 293
BX538160.1 - 3 310
HM991672.1 - 2 236

View Full Product Details