Gene Transcripts

Gene Symbol
GXYLT2
Entrez Gene ID
727936
Gene Name
glucoside xylosyltransferase 2
Gene Aliases
GLT8D4
Chromosome Location
Chr. 3: 72848522 - 72975374 on Build GRCh38
UniGene ID
Hs.710275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080393.1 NP_001073862.1 7 1172
XM_011534067.2 XP_011532369.1 7 1038
XM_011534068.1 XP_011532370.1 7 1057
XM_017007123.1 XP_016862612.1 6 789
XM_017007124.1 XP_016862613.1 6 918
GenBank mRNA BC127733.1 - 6 818

View Full Product Details