Gene Transcripts

Gene Symbol
SLC15A5
Entrez Gene ID
729025
Gene Name
solute carrier family 15 member 5
Gene Aliases
Chromosome Location
Chr. 12: 16188485 - 16277685 on Build GRCh38
UniGene ID
Hs.692712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170798.1 NP_001164269.1 3 708

View Full Product Details