Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135109.2 | NP_001128581.1 | 2 | 373 | |
NM_001206671.2 | NP_001193600.1 | 6 | 919 | ||
NM_001206672.2 | NP_001193601.1 | 4 | 676 | ||
NM_024557.4 | NP_078833.3 | 6 | 916 | ||
NR_045405.1 | 7 | 968 | |||
XM_006718317.3 | XP_006718380.1 | 6 | 1048 | ||
XM_006718318.3 | XP_006718381.1 | 6 | 961 | ||
XM_017018287.1 | XP_016873776.1 | 6 | 1048 | ||
XM_017018288.1 | XP_016873777.1 | 7 | 1045 | ||
XM_017018289.1 | XP_016873778.1 | 7 | 992 | ||
XM_017018290.1 | XP_016873779.1 | 5 | 815 | ||
GenBank mRNA | AK021670.1 | - | 5 | 940 | |
AK293933.1 | - | 6 | 966 | ||
AK296505.1 | - | 7 | 933 | ||
AK315379.1 | - | 6 | 875 | ||
AL832601.2 | - | 2 | 334 | ||
AM422214.1 | - | 4 | 622 | ||
AY326435.1 | - | 6 | 865 | ||
BC022455.1 | - | 6 | 898 | 4792930 | |
BX647514.1 | - | 3 | 516 | ||
DA186330.1 | - | 2 | 373 | ||
KU178773.1 | - | 6 | 862 | ||
KU178774.1 | - | 5 | 768 | ||
KU178775.1 | - | 2 | 319 |