Gene Transcripts

Gene Symbol
WDR26
Entrez Gene ID
80232
Gene Name
WD repeat domain 26
Gene Aliases
CDW2, GID7, MIP2
Chromosome Location
Chr. 1: 224385143 - 224434299 on Build GRCh38
UniGene ID
Hs.497873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115113.2 NP_001108585.2 9 1483
NM_025160.6 NP_079436.4 9 1531
XM_005273273.4 XP_005273330.1 9 1432
XM_006711817.3 XP_006711880.1 9 1429
XM_011544281.2 XP_011542583.1 9 1272
XM_011544282.2 XP_011542584.1 6 594
XM_017002396.1 XP_016857885.1 9 1120
GenBank mRNA AB209887.1 - 9 1531
AK024669.1 - 5 545
AK225061.1 - 5 545
AK299230.1 - 6 716
AY221751.1 - 9 1321
AY304473.1 - 9 1088
BC031471.2 - 4 474
BC034498.1 - 11 1523
BC052301.1 - 8 927 6420398
BC063817.1 - 5 593
EF011612.1 - 8 926

View Full Product Details