Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013431.2 | NP_038459.1 | 4 | 643 | |
GenBank mRNA | AF350018.1 | - | 4 | 461 | |
AF350019.1 | - | 4 | 461 | ||
AJ001683.1 | - | 4 | 643 | ||
BC017784.1 | - | 4 | 475 | 4619154 | |
U96845.1 | - | 4 | 493 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199805.1 | NP_001186734.1 | 4 | 322 | |
GenBank mRNA | AF461811.1 | - | 4 | 331 | |
AK292020.1 | - | 4 | 435 | ||
AK310717.1 | - | 4 | 435 | ||
X54870.1 | - | 1 | 98 |