Gene Transcripts

Gene Symbol
TLN2
Entrez Gene ID
83660
Gene Name
talin 2
Gene Aliases
ILWEQ
Chromosome Location
Chr. 15: 62390527 - 62844631 on Build GRCh38
UniGene ID
Hs.569438
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015059.2 NP_055874.2 47 6442
XM_005254708.4 XP_005254765.1 50 6810
XM_005254710.4 XP_005254767.1 51 6948
XM_005254711.4 XP_005254768.1 50 7170
XM_005254712.4 XP_005254769.1 50 6827
XM_005254713.4 XP_005254770.1 50 7724
XM_005254714.3 XP_005254771.1 49 6713
XM_005254715.2 XP_005254772.1 49 7419
XM_006720717.3 XP_006720780.1 50 7544
XM_017022665.1 XP_016878154.1 50 7895
XM_017022666.1 XP_016878155.1 50 6754
XM_017022667.1 XP_016878156.1 48 6616
XM_017022668.1 XP_016878157.1 50 7543
XM_017022669.1 XP_016878158.1 49 7409
GenBank mRNA AB002318.2 - 33 4616
AB621808.1 - 28 3725
AF402000.1 - 47 6442
BX647654.1 - 18 4356

View Full Product Details