Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001122979.2 | NP_001116451.1 | 7 | 1140 | |
NM_001170631.1 | NP_001164102.1 | 7 | 2025 | ||
NM_032029.4 | NP_114418.2 | 6 | 1223 | ||
XM_011510051.2 | XP_011508353.1 | 8 | 1974 | ||
XM_011510052.2 | XP_011508354.1 | 7 | 1735 | ||
XM_017002442.1 | XP_016857931.1 | 8 | 1970 | ||
GenBank mRNA | AK298394.1 | - | 7 | 2001 | |
AY063125.1 | - | 5 | 1408 | ||
AY063126.1 | - | 1 | 1109 | ||
BC109263.1 | - | 6 | 747 | ||
BC109264.2 | - | 6 | 747 |