Gene Transcripts

Gene Symbol
ARMC2
Entrez Gene ID
84071
Gene Name
armadillo repeat containing 2
Gene Aliases
bA787I22.1
Chromosome Location
Chr. 6: 108848410 - 108990738 on Build GRCh38
UniGene ID
Hs.645481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286609.1 NP_001273538.1 14 2224
NM_032131.5 NP_115507.4 15 2297
XM_005267154.3 XP_005267211.1 15 2578
XM_005267155.2 XP_005267212.1 15 2313
XM_005267157.3 XP_005267214.1 15 2221
XM_006715573.2 XP_006715636.1 16 2403
XM_006715574.2 XP_006715637.1 12 1733
XM_011536166.1 XP_011534468.1 16 2388
XM_011536167.1 XP_011534469.1 15 2174
XM_011536168.2 XP_011534470.1 15 2291
XM_011536170.2 XP_011534472.1 15 2291
XM_011536172.2 XP_011534474.1 14 2148
XM_017011350.1 XP_016866839.1 14 2226
XM_017011352.1 XP_016866841.1 14 2127
GenBank mRNA AK095234.1 - 14 2140
AK292499.1 - 15 2291
AK294574.1 - 14 2224
AL136754.1 - 11 1647
BC030603.2 - 15 2291 4824783

View Full Product Details