Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193452.1 | NP_001180381.1 | 3 | 689 | |
NM_032349.3 | NP_115725.1 | 3 | 619 | ||
XM_005255633.4 | XP_005255690.1 | 2 | 3691 | ||
GenBank mRNA | AK092642.1 | - | 2 | 1313 | |
BC006223.2 | - | 3 | 597 | 3944355 |