Gene Transcripts

Gene Symbol
RTN4IP1
Entrez Gene ID
84816
Gene Name
reticulon 4 interacting protein 1
Gene Aliases
NIMP, OPA10
Chromosome Location
Chr. 6: 106559237 - 106630921 on Build GRCh38
UniGene ID
Hs.155839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032730.5 NP_116119.2 1 612
XM_017011375.1 XP_016866864.1 1 612
XM_017011376.1 XP_016866865.1 1 612
GenBank mRNA AF336861.1 - 1 186
AF439711.1 - 1 135
AK313962.1 - 1 530
AY063761.1 - 1 135
BC006399.2 - 1 407 4309263
BQ936729.1 - 1 590

View Full Product Details