Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318834.1 | NP_001305763.1 | 8 | 643 | |
NM_032933.5 | NP_116322.3 | 7 | 528 | ||
NR_134875.1 | 7 | 528 | |||
NR_134876.1 | 7 | 565 | |||
NR_134877.1 | 7 | 528 | |||
XM_011526233.2 | XP_011524535.1 | 7 | 518 | ||
XM_017026043.1 | XP_016881532.1 | 7 | 518 | ||
XM_017026044.1 | XP_016881533.1 | 7 | 518 | ||
XM_017026045.1 | XP_016881534.1 | 6 | 442 | ||
XM_017026046.1 | XP_016881535.1 | 7 | 524 | ||
XM_017026047.1 | XP_016881536.1 | 7 | 448 | ||
XM_017026049.1 | XP_016881538.1 | 7 | 524 | ||
XM_017026050.1 | XP_016881539.1 | 7 | 546 | ||
XM_017026051.1 | XP_016881540.1 | 7 | 448 | ||
GenBank mRNA | AB253738.1 | - | 7 | 428 | |
AB253739.1 | - | 8 | 543 | ||
AK123623.1 | - | 7 | 555 | ||
AK126247.1 | - | 7 | 520 | ||
AK294916.1 | - | 7 | 524 | ||
AK303553.1 | - | 7 | 412 | ||
BC054507.1 | - | 1 | 25 | ||
BC114510.1 | - | 7 | 464 | 40023922 | |
BC114561.1 | - | 7 | 464 | 40023923 | |
DA385968.1 | - | 6 | 443 |