Gene Transcripts

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
Chromosome Location
Chr. 11: 6621324 - 6655849 on Build GRCh38
UniGene ID
Hs.199850
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003737.3 NP_003728.1 5 2774
GenBank mRNA AB053446.1 - 5 2777

View Full Product Details