Gene Transcripts

Gene Symbol
URB2
Entrez Gene ID
9816
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Gene Aliases
KIAA0133, NET10, NPA2
Chromosome Location
Chr. 1: 229626216 - 229660200 on Build GRCh38
UniGene ID
Hs.533628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001314021.1 NP_001300950.1 2 242
NM_014777.3 NP_055592.2 2 211
XM_005273360.3 XP_005273417.1 2 180
GenBank mRNA AK291013.1 - 2 211
BC059381.1 - 2 186
BC114559.1 - 2 164 40082897
BC114963.1 - 2 133 40082900
D50923.2 - 2 193
DC369323.1 - 2 211

View Full Product Details