Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
SLC12A6 | NM_001042494.1 | NP_001035959.1 | ||
NM_001042495.1 | NP_001035960.1 | |||
NM_001042496.1 | NP_001035961.1 | |||
NM_001042497.1 | NP_001035962.1 | |||
NM_005135.2 | NP_005126.1 | |||
NM_133647.1 | NP_598408.1 | |||
XM_006720793.3 | XP_006720856.1 | |||
AF105366.1 | AAD39742.1 | |||
AF108831.1 | AAD25337.1 | |||
AF116242.1 | AAF24986.1 | |||
AF477977.1 | AAL85335.1 | |||
AF531258.1 | AAQ10026.1 | |||
AF531259.1 | AAQ10027.1 | |||
AF531260.1 | AAQ10028.1 | |||
AK128133.1 | ||||
AK292550.1 | ||||
AK315283.1 | ||||
AL117500.1 | CAB55965.1 | |||
BC051744.1 | AAH51744.1 | |||
BC070107.1 | ||||
BC126241.1 | ||||
BC126243.1 | ||||
BC150584.1 | ||||
BX648195.1 | CAI46042.1 | |||
DQ138323.1 | ABA02873.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1052620 | Chr15:32612402 - 34538731 on Build GRCh38 | Gain | CHRM5 FMN1 NOP10 ARHGAP11A LOC105370757 LOC101928042 KATNBL1 MIR1233-1 EMC7 SCG5 LOC100131315 PGBD4 GOLGA8B EMC4 RYR3 MIR1233-2 GREM1 LPCAT4 TMCO5B SLC12A6 GOLGA8A LOC101928134 AVEN NUTM1 |
esv33337 | Chr15:31689863 - 34575097 on Build GRCh38 | Gain+Loss | ULK4P1 CHRM5 FMN1 LOC101928042 EMC7 LOC101060588 SCG5 WHAMMP1 PGBD4 GOLGA8N GOLGA8B EMC4 RYR3 LOC100653133 MIR1233-2 GREM1 CHRNA7 SLC12A6 GOLGA8A AVEN GOLGA8K LOC100996255 NOP10 ARHGAP11A LOC105370940 LOC105370757 GOLGA8O KATNBL1 MIR1233-1 LOC100131315 LPCAT4 TMCO5B LOC101928134 NUTM1 |
esv3569431 | Chr15:33564570 - 35080307 on Build GRCh38 | Gain | ACTC1 CHRM5 NOP10 KATNBL1 GJD2 ZNF770 MIR1233-1 EMC7 PGBD4 GOLGA8B EMC4 RYR3 LOC101928174 MIR1233-2 LPCAT4 SLC12A6 AQR GOLGA8A AVEN NUTM1 |
dgv4456n54 | Chr15:32604149 - 34557061 on Build GRCh38 | Gain | CHRM5 FMN1 LOC101928042 EMC7 SCG5 PGBD4 GOLGA8N GOLGA8B EMC4 RYR3 MIR1233-2 GREM1 SLC12A6 GOLGA8A AVEN NOP10 ARHGAP11A LOC105370757 KATNBL1 MIR1233-1 LOC100131315 LPCAT4 TMCO5B LOC101928134 NUTM1 |